Anti-OLIG1 Rabbit Monoclonal Antibody [clone: EPR6790]

Toimittaja: Abcam
ABCAAB124908-100EA 1050 EUR
ABCAAB124908-100
Anti-OLIG1 Rabbit Monoclonal Antibody [clone: EPR6790]
Vasta-aineet
Type: Primary
Antigen: OLIG1 (oligodendrocyte transcription factor 1)
Clonality: monoclonal
Clone: EPR6790
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR