Anti-SUN2 Rabbit Monoclonal Antibody [clone: EPR6557]

Toimittaja: Abcam
ABCAAB124916-100EA 1020 EUR
ABCAAB124916-100
Anti-SUN2 Rabbit Monoclonal Antibody [clone: EPR6557]
Vasta-aineet
Type: Primary
Antigen: SUN2 (Sad1 and UNC84 domain containing 2)
Clonality: monoclonal
Clone: EPR6557
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR