Anti-Mib1/Mindbomb Rabbit Monoclonal Antibody [clone: EPR2762]

Toimittaja: Abcam
ABCAAB124929-100EA 1050 EUR
ABCAAB124929-100
Anti-Mib1/Mindbomb Rabbit Monoclonal Antibody [clone: EPR2762]
Vasta-aineet
Type: Primary
Antigen: Mib1/Mindbomb
Clonality: monoclonal
Clone: EPR2762
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR