Anti-UCHL5 Rabbit Monoclonal Antibody [clone: EPR4896]

Toimittaja: Abcam
ABCAAB124931-100EA 1010 EUR
ABCAAB124931-100
Anti-UCHL5 Rabbit Monoclonal Antibody [clone: EPR4896]
Vasta-aineet
Type: Primary
Antigen: UCHL5 (ubiquitin carboxyl-terminal hydrolase L5)
Clonality: monoclonal
Clone: EPR4896
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR