Anti-Dishevelled 2 Rabbit monoclonal antibody unconjugated [clone: EPNCIR145]

Toimittaja: Abcam
ABCAAB124933-100EA 935 EUR
ABCAAB124933-100
Anti-Dishevelled 2 Rabbit monoclonal antibody unconjugated [clone: EPNCIR145]
Vasta-aineet
Type: Primary
Antigen: Dishevelled 2
Clonality: monoclonal
Clone: EPNCIR145
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR