Anti-MECOM Rabbit Monoclonal Antibody [clone: EPR5996]

Toimittaja: Abcam
ABCAAB124934-100EA 1010 EUR
ABCAAB124934-100
Anti-MECOM Rabbit Monoclonal Antibody [clone: EPR5996]
Vasta-aineet
Type: Primary
Antigen: MECOM (MDS1 and EVI1 complex locus)
Clonality: monoclonal
Clone: EPR5996
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR