Anti-ERN1 Rabbit Monoclonal Antibody [clone: EPR5253]

Toimittaja: Abcam
ABCAAB124945-100EA 1120 EUR
ABCAAB124945-100
Anti-ERN1 Rabbit Monoclonal Antibody [clone: EPR5253]
Vasta-aineet
Type: Primary
Antigen: ERN1 (endoplasmic reticulum to nucleus signaling 1)
Clonality: monoclonal
Clone: EPR5253
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR