Anti-RAP1A G12V Rabbit Monoclonal Antibody [clone: EP878]

Toimittaja: Abcam
ABCAAB124963-100EA 904 EUR
ABCAAB124963-100
Anti-RAP1A G12V Rabbit Monoclonal Antibody [clone: EP878]
Vasta-aineet
Type: Primary
Antigen: RAP1A G12V
Clonality: monoclonal
Clone: EP878
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR