Anti-ATP7B Rabbit Monoclonal Antibody [clone: EPR6794]

Toimittaja: Abcam
ABCAAB124973-100EA 1030 EUR
ABCAAB124973-100
Anti-ATP7B Rabbit Monoclonal Antibody [clone: EPR6794]
Vasta-aineet
Type: Primary
Antigen: ATP7B (ATPase, Cu++ transporting, beta polypeptide)
Clonality: monoclonal
Clone: EPR6794
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR