Anti-HDAC2 Rabbit monoclonal antibody unconjugated [clone: EPR5001]

Toimittaja: Abcam
ABCAAB124974-100EA 1150 EUR
ABCAAB124974-100
Anti-HDAC2 Rabbit monoclonal antibody unconjugated [clone: EPR5001]
Vasta-aineet
Type: Primary
Antigen: HDAC2 (histone deacetylase 2)
Clonality: monoclonal
Clone: EPR5001
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR