Anti-CFL1 Rabbit monoclonal antibody unconjugated [clone: EPR6375]

Toimittaja: Abcam
ABCAAB124979-100EA 1020 EUR
ABCAAB124979-100
Anti-CFL1 Rabbit monoclonal antibody unconjugated [clone: EPR6375]
Vasta-aineet
Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: monoclonal
Clone: EPR6375
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR