Anti-Neurokinin A Receptor Rabbit Monoclonal Antibody [clone: EPR6838]

Toimittaja: Abcam
ABCAAB124998-100EA 883 EUR
ABCAAB124998-100
Anti-Neurokinin A Receptor Rabbit Monoclonal Antibody [clone: EPR6838]
Vasta-aineet
Type: Primary
Antigen: Neurokinin A Receptor
Clonality: monoclonal
Clone: EPR6838
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR