Anti-EIF3E Rabbit monoclonal antibody unconjugated [clone: EPR6877]

Toimittaja: Abcam
ABCAAB125033-100EA 935 EUR
ABCAAB125033-100
Anti-EIF3E Rabbit monoclonal antibody unconjugated [clone: EPR6877]
Vasta-aineet
Type: Primary
Antigen: EIF3E (eukaryotic translation initiation factor 3, subunit E)
Clonality: monoclonal
Clone: EPR6877
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR