Anti-GBA Rabbit monoclonal antibody unconjugated [clone: EPR5142]

Toimittaja: Abcam
ABCAAB125065-100EA 1120 EUR
ABCAAB125065-100
Anti-GBA Rabbit monoclonal antibody unconjugated [clone: EPR5142]
Vasta-aineet
Type: Primary
Antigen: GBA (glucosidase, beta, acid)
Clonality: monoclonal
Clone: EPR5142
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR