Anti-Band 3 Rabbit monoclonal antibody unconjugated [clone: EPR1424]

Toimittaja: Abcam
ABCAAB125070-100EA 926 EUR
ABCAAB125070-100
Anti-Band 3 Rabbit monoclonal antibody unconjugated [clone: EPR1424]
Vasta-aineet
Type: Primary
Antigen: Band 3
Clonality: monoclonal
Clone: EPR1424
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR