Anti-mCherry Mouse Monoclonal Antibody [clone: 1C51]

Toimittaja: Abcam
ABCAAB125096-100EA 1040 EUR
ABCAAB125096-100
Anti-mCherry Mouse Monoclonal Antibody [clone: 1C51]
Vasta-aineet
Type: Primary
Antigen: mCherry
Clonality: monoclonal
Clone: 1C51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR