Anti-Creatine kinase B type Mouse monoclonal antibody unconjugated [clone: CK-BYK/21E10]

Toimittaja: Abcam
ABCAAB125114-50EA 1110 EUR
ABCAAB125114-50
Anti-Creatine kinase B type Mouse monoclonal antibody unconjugated [clone: CK-BYK/21E10]
Vasta-aineet
Type: Primary
Antigen: Creatine kinase B type
Clonality: monoclonal
Clone: CK-BYK/21E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR