Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 1D5]

Toimittaja: Abcam
ABCAAB125687-100EA 1050 EUR
ABCAAB125687-100
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 1D5]
Vasta-aineet
Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: monoclonal
Clone: 1D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR