Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abcam
ABCAAB125856-500EA 467 EUR
ABCAAB125856-500
Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Type: Secondary
Antigen: Turkey IgY
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR