Anti-IgG Rabbit Monoclonal Antibody

Toimittaja: Abcam
ABCAAB125900-100EA 833 EUR
ABCAAB125900-100
Anti-IgG Rabbit Monoclonal Antibody
Vasta-aineet
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR