Anti-SLC4A8 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB125910-50EA 992 EUR
ABCAAB125910-50
Anti-SLC4A8 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SLC4A8 (solute carrier family 4, sodium bicarbonate cotransporter, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR