Anti-RAD23B Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB125918-50EA 997 EUR
ABCAAB125918-50
Anti-RAD23B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RAD23B (RAD23 homolog B (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR