Anti-TIMM17A Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126044-50EA 992 EUR
ABCAAB126044-50
Anti-TIMM17A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TIMM17A (translocase of inner mitochondrial membrane 17 homolog A (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR