Anti-TMED7 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126094-50EA 946 EUR
ABCAAB126094-50
Anti-TMED7 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TMED7 (transmembrane emp24 protein transport domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR