Anti-SMARCC1 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126180-50EA 1030 EUR
ABCAAB126180-50
Anti-SMARCC1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SMARCC1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR