Anti-ATP5L Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB126181-50EA 955 EUR
ABCAAB126181-50
Anti-ATP5L Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: ATP5L (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR