Anti-BFSP1 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB126235-50EA 946 EUR
ABCAAB126235-50
Anti-BFSP1 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: BFSP1 (beaded filament structural protein 1, filensin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR