Anti-RPP25 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126338-100EA 969 EUR
ABCAAB126338-100
Anti-RPP25 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RPP25 (ribonuclease P/MRP 25kDa subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR