Anti-C9orf142 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB126353-100EA 1010 EUR
ABCAAB126353-100
Anti-C9orf142 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: C9orf142 (chromosome 9 open reading frame 142)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR