Anti-TMEM229B Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126378-100EA 969 EUR
ABCAAB126378-100
Anti-TMEM229B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TMEM229B (transmembrane protein 229B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR