Anti-VMO1 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126510-100EA 969 EUR
ABCAAB126510-100
Anti-VMO1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: VMO1 (vitelline membrane outer layer 1 homolog (chicken))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR