Anti-ETFDH Mouse monoclonal antibody unconjugated [clone: 3H2BG1]

Toimittaja: Abcam
ABCAAB126576-100EA 1020 EUR
ABCAAB126576-100
Anti-ETFDH Mouse monoclonal antibody unconjugated [clone: 3H2BG1]
Vasta-aineet
Type: Primary
Antigen: ETFDH (electron-transferring-flavoprotein dehydrogenase)
Clonality: monoclonal
Clone: 3H2BG1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR