Anti-MAPK10 Rabbit Monoclonal Antibody [clone: EPR5493]

Toimittaja: Abcam
ABCAAB126591-100EA 933 EUR
ABCAAB126591-100
Anti-MAPK10 Rabbit Monoclonal Antibody [clone: EPR5493]
Vasta-aineet
Type: Primary
Antigen: MAPK10 (mitogen-activated protein kinase 10)
Clonality: monoclonal
Clone: EPR5493
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR