Anti-SERPINC1 Rabbit Monoclonal Antibody [clone: EP5372]

Toimittaja: Abcam
ABCAAB126598-100EA 1020 EUR
ABCAAB126598-100
Anti-SERPINC1 Rabbit Monoclonal Antibody [clone: EP5372]
Vasta-aineet
Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: monoclonal
Clone: EP5372
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR