Anti-SFRP1 Rabbit Monoclonal Antibody [clone: EPR7003]

Toimittaja: Abcam
ABCAAB126613-100EA 1050 EUR
ABCAAB126613-100
Anti-SFRP1 Rabbit Monoclonal Antibody [clone: EPR7003]
Vasta-aineet
Type: Primary
Antigen: SFRP1 (secreted frizzled-related protein 1)
Clonality: monoclonal
Clone: EPR7003
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR