Anti-ME2 Rabbit Monoclonal Antibody [clone: EPR7218]

Toimittaja: Abcam
ABCAAB126616-100EA 1020 EUR
ABCAAB126616-100
Anti-ME2 Rabbit Monoclonal Antibody [clone: EPR7218]
Vasta-aineet
Type: Primary
Antigen: ME2 (malic enzyme 2, NAD(+)-dependent, mitochondrial)
Clonality: monoclonal
Clone: EPR7218
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR