Anti-S1PR3 Rabbit Monoclonal Antibody [clone: EPR4541]

Toimittaja: Abcam
ABCAAB126622-100EA 1030 EUR
ABCAAB126622-100
Anti-S1PR3 Rabbit Monoclonal Antibody [clone: EPR4541]
Vasta-aineet
Type: Primary
Antigen: S1PR3 (sphingosine-1-phosphate receptor 3)
Clonality: monoclonal
Clone: EPR4541
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR