Anti-FKBP5 Rabbit monoclonal antibody unconjugated [clone: EPR6617]

Toimittaja: Abcam
ABCAAB126715-100EA 1020 EUR
ABCAAB126715-100
Anti-FKBP5 Rabbit monoclonal antibody unconjugated [clone: EPR6617]
Vasta-aineet
Type: Primary
Antigen: FKBP5 (FK506 binding protein 5)
Clonality: monoclonal
Clone: EPR6617
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR