Anti-MCM3 Rabbit Monoclonal Antibody [clone: EPR7081]

Toimittaja: Abcam
ABCAAB126723-100EA 1020 EUR
ABCAAB126723-100
Anti-MCM3 Rabbit Monoclonal Antibody [clone: EPR7081]
Vasta-aineet
Type: Primary
Antigen: MCM3 (minichromosome maintenance complex component 3)
Clonality: monoclonal
Clone: EPR7081
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR