Anti-CLIC2 Rabbit Monoclonal Antibody [clone: EPR6495]

Toimittaja: Abcam
ABCAAB126727-100EA 900 EUR
ABCAAB126727-100
Anti-CLIC2 Rabbit Monoclonal Antibody [clone: EPR6495]
Vasta-aineet
Type: Primary
Antigen: CLIC2 (chloride intracellular channel 2)
Clonality: monoclonal
Clone: EPR6495
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR