Anti-EIF5A2 Rabbit monoclonal antibody unconjugated [clone: EPR7411]

Toimittaja: Abcam
ABCAAB126735-100EA 935 EUR
ABCAAB126735-100
Anti-EIF5A2 Rabbit monoclonal antibody unconjugated [clone: EPR7411]
Vasta-aineet
Type: Primary
Antigen: EIF5A2 (eukaryotic translation initiation factor 5A2)
Clonality: monoclonal
Clone: EPR7411
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR