Anti-intestinal FABP Rabbit Monoclonal Antibody [clone: EPR7137]

Toimittaja: Abcam
ABCAAB126744-100EA 925 EUR
ABCAAB126744-100
Anti-intestinal FABP Rabbit Monoclonal Antibody [clone: EPR7137]
Vasta-aineet
Type: Primary
Antigen: intestinal FABP
Clonality: monoclonal
Clone: EPR7137
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR