Anti-CD163L1 Rabbit Monoclonal Antibody [clone: EPR6539]

Toimittaja: Abcam
ABCAAB126756-100EA 1020 EUR
ABCAAB126756-100
Anti-CD163L1 Rabbit Monoclonal Antibody [clone: EPR6539]
Vasta-aineet
Type: Primary
Antigen: CD163L1 (CD163 molecule-like 1)
Clonality: monoclonal
Clone: EPR6539
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR