Anti-UBE3A Rabbit Monoclonal Antibody [clone: EPR7330]

Toimittaja: Abcam
ABCAAB126765-100EA 923 EUR
ABCAAB126765-100
Anti-UBE3A Rabbit Monoclonal Antibody [clone: EPR7330]
Vasta-aineet
Type: Primary
Antigen: UBE3A (ubiquitin protein ligase E3A)
Clonality: monoclonal
Clone: EPR7330
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR