Anti-BMI1 Rabbit Monoclonal Antibody [clone: EPR3745]

Toimittaja: Abcam
ABCAAB126783-100EA 1130 EUR
ABCAAB126783-100
Anti-BMI1 Rabbit Monoclonal Antibody [clone: EPR3745]
Vasta-aineet
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: monoclonal
Clone: EPR3745
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR