Anti-DPYSL3 Rabbit Monoclonal Antibody [clone: EPR7593]

Toimittaja: Abcam
ABCAAB126787-100EA 1060 EUR
ABCAAB126787-100
Anti-DPYSL3 Rabbit Monoclonal Antibody [clone: EPR7593]
Vasta-aineet
Type: Primary
Antigen: DPYSL3 (dihydropyrimidinase-like 3)
Clonality: monoclonal
Clone: EPR7593
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR