Anti-CAMK2B Rabbit Monoclonal Antibody [clone: EPR6685]

Toimittaja: Abcam
ABCAAB126789-100EA 1020 EUR
ABCAAB126789-100
Anti-CAMK2B Rabbit Monoclonal Antibody [clone: EPR6685]
Vasta-aineet
Type: Primary
Antigen: CAMK2B (calcium/calmodulin-dependent protein kinase II beta)
Clonality: monoclonal
Clone: EPR6685
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR