Anti-QK1 Rabbit Monoclonal Antibody [clone: EPR7305]

Toimittaja: Abcam
ABCAAB126791-100EA 949 EUR
ABCAAB126791-100
Anti-QK1 Rabbit Monoclonal Antibody [clone: EPR7305]
Vasta-aineet
Type: Primary
Antigen: QK1
Clonality: monoclonal
Clone: EPR7305
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR