Anti-BCL7C Rabbit polyclonal antibody unconjugated
Toimittaja: Abcam
ABCAAB126944-50EA 946 EUR
ABCAAB126944-50
Anti-BCL7C Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: BCL7C (B-cell CLL/lymphoma 7C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human