Anti-NMUR1 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB126961-50EA 946 EUR
ABCAAB126961-50
Anti-NMUR1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: NMUR1 (neuromedin U receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR